missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEU-1/Sialidase-1 Antibody (OTI3B3), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
£195.00 - £417.00
Specifications
| Antigen | NEU-1/Sialidase-1 |
|---|---|
| Clone | OTI3B3 |
| Dilution | Western Blot 1:2000, Immunohistochemistry 1:150 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306330
|
Novus Biologicals
NBP3-44875-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306366
|
Novus Biologicals
NBP3-44875-30ul |
30 μL |
£195.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NEU-1/Sialidase-1 Monoclonal antibody specifically detects NEU-1/Sialidase-1 in Human samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| NEU-1/Sialidase-1 | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Acetylneuraminyl hydrolase, EC 3.2.1.18, exo-alpha-sialidase, FLJ93471, G9 sialidase, Lysosomal sialidase, N-acetyl-alpha-neuraminidase 1, NANH, NEU, SIAL1, sialidase 1 (lysosomal sialidase), sialidase-1 | |
| Human recombinant protein fragment corresponding to amino acids 48-315 of human NEU1 (NP_000425)produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| OTI3B3 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| metabolism | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 4758 | |
| IgG2a | |
| >95% | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title