missing translation for 'onlineSavingsMsg'
Learn More

Netupitant, MedChemExpress

Product Code. 16499577
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499577

Brand: Medchem Express HY16346

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Netupitant (CID-6451149) is a highly potent, selective and orally active neurokinin-1 (NK1) receptor antagonist with a Ki of 0.95 nM for hNK1 in CHO cells. Netupitant has antiemetic affect.

Netupitant (CID-6451149) is a highly potent, selective and orally active neurokinin-1 (NK1) receptor antagonist with a Ki of 0.95 nM for hNK1 in CHO cells. Netupitant has antiemetic affect.

Specifications

Chemical Name or Material Netupitant
CAS 290297-26-6
Color White
Purity Grade Notes Research
Molecular Formula C30H32F6N4O
Quantity 5 mg
Synonym CID 6451149
Solubility Information DMSO : 9.09 mg/mL (15.71 mM; ultrasonic and warming and heat to 60°C)
SMILES CN1CCN(C2=CC(C3=C(C)C=CC=C3)=C(N(C)C(C(C4=CC(C(F)(F)F)=CC(C(F)(F)F)=C4)(C)C)=O)C=N2)CC1
Molecular Weight (g/mol) 578.59
Formula Weight 578.59
Percent Purity 98.96%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.