missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Netrin G2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49653
This item is not returnable.
View return policy
Description
Netrin G2 Polyclonal antibody specifically detects Netrin G2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Netrin G2 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| bA479K20.1, KIAA0625, KIAA1857bA479K20.1 (novel protein), laminet 2, laminet-2, LHLL9381, LMNT2, MGC21884, netrin G1, netrin G2, netrin-G2, NTNG1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KEEEGLATYWQSITWSRYPSPLEANITLSWNKTVELTDDVVMTFEYG | |
| 0.1 mL | |
| Neuroscience | |
| 84628 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction