missing translation for 'onlineSavingsMsg'
Learn More

Netrin G2 Antibody (4F11), Novus Biologicals™

Product Code. 18432989 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432989 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432989 Supplier Novus Biologicals Supplier No. H00084628M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Netrin G2 Monoclonal antibody specifically detects Netrin G2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Netrin G2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4F11
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_115925
Gene Alias bA479K20.1, KIAA0625, KIAA1857bA479K20.1 (novel protein), laminet 2, laminet-2, LHLL9381, LMNT2, MGC21884, netrin G1, netrin G2, netrin-G2, NTNG1
Host Species Mouse
Immunogen NTNG2 (NP_115925, 21 a.a. ∽ 119 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ICKSWVTTDEGPTWEFYACQPKVMRLKDYVKVKVEPSGITCGDPPERFCSHENPYLCSNECDASNPDLAHPPRLMFDKEEEGLATYWQSITWSRYPSPL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 84628
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.