missing translation for 'onlineSavingsMsg'
Learn More

Netrin-4 Antibody (299612) [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30735506 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735506 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735506 Supplier Novus Biologicals Supplier No. FAB1132MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Netrin-4 Monoclonal antibody specifically detects Netrin-4 in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Netrin-4
Applications Western Blot
Classification Monoclonal
Clone 299612
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias beta-netrin, hepar-derived netrin-like protein, netrin 4, NTN4, PRO3091
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Netrin-4, Gly19-Val628, Accession # Q9JI33
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59277
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.