missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NETO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572701
This item is not returnable.
View return policy
Description
NETO1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. A similar gene in mice encodes a protein that plays a critical role in spatial learning and memory by regulating the function of synaptic N-methyl-D-aspartic acid receptor complexes in the hippocampus. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| NETO1 | |
| Polyclonal | |
| Unconjugated | |
| NETO1 | |
| AI851453; BCTL1; brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor class A domains protein 1; Btcl1; C130005O10Rik; NETO1; neuopilin- and tolloid-like 1; neuropilin (NRP) and tolloid (TLL)-like 1; neuropilin and tolloid like 1; neuropilin- and tolloid-like protein 1; neuropilin and tolloid-like protein 1; RGD1566269; si:dkey-82k12.12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 246317, 307206, 81832 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8R4I7, Q8TDF5 | |
| NETO1 | |
| NETO1 antibody for an 18 amino acid synthetic peptide near the carboxy terminus of human NETO1. The immunogen is within amino acids 350 - 400 of NETO1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction