missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NETO1 Monoclonal Antibody (532509)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524234
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. A similar gene in mice encodes a protein that plays a critical role in spatial learning and memory by regulating the function of synaptic N-methyl-D-aspartic acid receptor complexes in the hippocampus. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| NETO1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8TDF5 | |
| NETO1 | |
| CHO-derived recombinant human NETO1 isoform 3. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 532509 | |
| Unconjugated | |
| NETO1 | |
| AI851453; BCTL1; brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor class A domains protein 1; Btcl1; C130005O10Rik; NETO1; neuopilin- and tolloid-like 1; neuropilin (NRP) and tolloid (TLL)-like 1; neuropilin and tolloid like 1; neuropilin- and tolloid-like protein 1; neuropilin and tolloid-like protein 1; RGD1566269; si:dkey-82k12.12 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 81832 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction