missing translation for 'onlineSavingsMsg'
Learn More

NETO1 Antibody, FITC, Novus Biologicals™

Product Code. 18369434 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369434 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369434 Supplier Novus Biologicals Supplier No. NBP299822F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NETO1 Polyclonal specifically detects NETO1 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NETO1
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor classA domains protein 1, BTCL1BCTL1neuropilin and tolloid-like protein 1, FLJ41325, neuropilin (NRP) and tolloid (TLL)-like 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant NETO1 (NP_620416.1; Met 1-Thr 344).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 81832
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.