missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NETO1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99822R
This item is not returnable.
View return policy
Description
NETO1 Polyclonal specifically detects NETO1 in Human samples. It is validated for ELISA.
Specifications
| NETO1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| DyLight 550 | |
| Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor classA domains protein 1, BTCL1BCTL1neuropilin and tolloid-like protein 1, FLJ41325, neuropilin (NRP) and tolloid (TLL)-like 1 | |
| Produced in rabbits immunized with purified, recombinant NETO1 (NP_620416.1; Met 1-Thr 344). | |
| 0.1 mL | |
| Vision | |
| 81832 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction