missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NET1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528411
This item is not returnable.
View return policy
Description
Recommended positive controls: NET1 tansfected 293T cell. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Acts as guanine nucleotide exchange factor (GEF) for RhoA GTPase. May be involved in activation of the SAPK/JNK pathway.
Specifications
| NET1 | |
| Polyclonal | |
| Unconjugated | |
| NET1 | |
| Arhgef8; guanine nucleotide regulatory protein (oncogene); mNET1; NET1; Net1 homolog; Net1a; neuroepithelial cell transforming 1; neuroepithelial cell transforming gene 1; neuroepithelial cell-transforming gene 1 protein; neuroepithelioma transforming gene 1; p65 Net1 proto-oncogene protein; Proto-oncogene p65 Net1; Rho guanine nucleotide exchange factor (GEF) 8; rho guanine nucleotide exchange factor 8; RP11-116G8.1; small GTP-binding protein regulator | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10276 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.75 mg/mL | |
| PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q7Z628 | |
| NET1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human NET1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction