Learn More
NET1 Antibody (CL3063) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-43818
Product Code. 30296547
  • £458.00 / 100µL
Description
NET1 Monoclonal antibody specifically detects NET1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry (Paraffin),Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NET1
Monoclonal
LYOPH
Immunohistochemistry-Paraffin 1:200 - 1:500
ARHGEF8neuroepithelial cell-transforming gene 1 protein, guanine nucleotide regulatory protein (oncogene), NET1A, neuroepithelial cell transforming 1, neuroepithelial cell transforming gene 1, neuroepithelioma transforming gene 1, p65 Net1 proto-oncogene protein, Proto-oncogene p65 Net1, Rho guanine nucleotide exchange factor (GEF) 8, Rho guanine nucleotide exchange factor 8, small GTP-binding protein regulator
This antibody was generated using a recombinant protein sequence of P23975, with the exact immunogen sequence remaining proprietary.
100 μL
Primary
Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL.
Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles.
IgG1
Immunohistochemistry (Paraffin), Immunohistochemistry
CL3063
Unconjugated
Lyophilized in 5% Trehalose.
Mouse
Protein A purified
RUO
10276
Human, Mouse, Rat
Purified