Learn More
SPEX Certiprep™ Nesting Tray Set
Product Code. 15350422 Shop All SPEX Certiprep Products
Product Code. 15350422
Brand: SPEX Certiprep™ 1690T
This item requires a price update prior to being ordered.
Request a Price for this item here.
Keeps the samples secure during the grinding process.
  • Tray that allows titre plates to be held securely and stacked vertically on the 1690 adjustable clamp as used with the MiniG homogeniser from SPEX SamplePrep.
  • Tray to hold up two stacked deep well plates in 1690 clamp
Specifications
For Use With (Application) Accessory for the SPEX MiniG homogeniser