missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nestin Mouse, anti-Human, Unconjugated, Clone: 10C2, STEMCELL Technologies
Mouse Monoclonal Antibody
£187.65 - £441.45
Specifications
| Antigen | Nestin |
|---|---|
| Clone | 10C2 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17198961
|
Stemcell Technologies
60091.1 |
25 μg |
£187.65
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17188961
|
Stemcell Technologies
60091 |
100 μg |
£441.45
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The 10C2 antibody reacts with human nestin, a class VI intermediate filament (IF) protein expressed by embryonic neuroepithelial stem cells in the developing nervous system and by pancreatic islet and mesenchymal progenitor cells
- Nestin expression is also observed in glioblastomas and many primary and metastatic melanomas and it is a useful angiogenic marker to evaluate neovascularity of endothelial cells in tumors
- Nestin is required for the survival and renewal of neural progenitor cells and plays roles in cellular remodeling by forming heterodimers with vimentin or alpha-internexin, a process that appears to be tightly regulated by phosphorylation
- Nestin localizes to the growth cones during axon elongation in differentiating neurons and may play a role in growth cone guidance
- Nestin expression is down-regulated during differentiation, typically disappearing by embryonic stage E18
- The 10C2 antibody reportedly cross-reacts weakly with monkey nestin but not rodent nestin
Specifications
| Nestin | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Cynomolgus, Human | |
| Primary |
| 10C2 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title