missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nestin Monoclonal Antibody (196908), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA523650
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Nestin Monoclonal antibody specifically detects Nestin in Human samples. It is validated for Flow Cytometry.Specifications
Nestin | |
Monoclonal | |
APC | |
Nes | |
AA166324; C78523; ESTM46; FLJ21841; intermediate filament protein; Nbla00170; Nes; Nestin | |
Mouse | |
Protein A/G | |
RUO | |
10763 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
196908 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
P48681 | |
Nes | |
NS0 mouse myeloma cell line transfected with human Nestin | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |