missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nestin Antibody (10C2) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80869
This item is not returnable.
View return policy
Description
Nestin Monoclonal specifically detects Nestin in Human, Mouse (Negative), Rabbit (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunocytochemistry, CyTOF-ready.
Specifications
| Nestin | |
| Monoclonal | |
| 2.4 mg/mL | |
| Western Blot : 1:1000 - 1:5000, Flow Cytometry : 1 ug per million cells, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence : 1:50 - 1:200, Immunohistochemistry-Paraffin : 1:100 - 1:200, Immunohistochemistry-Frozen : 1:100 - 1:200, Immunocytochemistry, CyTOF-ready | |
| FLJ21841, nestin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| It does not cross-react mouse, rat or rabbit. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF | |
| 10C2 | |
| Unconjugated | |
| PBS | |
| NES | |
| A fragment of the human Nestin protein corresponding to residues 1464-1614. [UniProt P48681] | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cancer Stem Cells, Cell Biology, Cellular Markers, Extracellular Matrix, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
| 10763 | |
| Human, Mouse (Negative), Rabbit (Negative), Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction