missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NESS 0327
GREENER_CHOICE

Product Code. 18727646
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18727646

Brand: Tocris Bioscience™ 5746/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ultra-high affinity and selective CB1 antagonist

NESS 0327 is an ultra-high affinity and selective CB1 receptor antagonist (Ki = 350 fM). Exhibits >60-fold selectivity for CB1 over CB2 receptors. Antagonizes the inhibitory effects of WIN 55,212-2 (Cat. No. 1038) on electrically evoked contractions in mouse isolated vas deferens ex vivo. Also antagonizes the antinociceptive effect of WIN 55,212-2 in tail-flick and hot plate tests in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Chloro-1-(2,4-dichlorophenyl)-1,4,5,6-tetrahydro-N-1-piperidinyl-benzo[6,7 ]cyclohepta[1,2-c]pyrazole-3-carboxamide
CAS 494844-07-4
Quantity 50 mg
Target Cannabinoid R1/CB1 Antagonists
Molecular Formula C24H23Cl3N4O
Formula Weight 7676
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.