missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NESS 0327

Description
NESS 0327 is an ultra-high affinity and selective CB1 receptor antagonist (Ki = 350 fM). Exhibits >60-fold selectivity for CB1 over CB2 receptors. Antagonizes the inhibitory effects of WIN 55,212-2 (Cat. No. 1038) on electrically evoked contractions in mouse isolated vas deferens ex vivo. Also antagonizes the antinociceptive effect of WIN 55,212-2 in tail-flick and hot plate tests in vivo.
Specifications
Specifications
| Chemical Name or Material | 8-Chloro-1-(2,4-dichlorophenyl)-1,4,5,6-tetrahydro-N-1-piperidinyl-benzo[6,7 ]cyclohepta[1,2-c]pyrazole-3-carboxamide |
| CAS | 494844-07-4 |
| Quantity | 10 mg |
| Target | Cannabinoid R1/CB1 Antagonists |
| Molecular Formula | C24H23Cl3N4O |
| Formula Weight | 1970 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?