missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nesprin 2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA578438

Product Code. 16357134

  • £484.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Control: NIH-3T3 Predicted Reactivity: Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a nuclear outer membrane protein that binds cytoplasmic F-actin. This binding tethers the nucleus to the cytoskeleton and aids in the maintenance of the structural integrity of the nucleus. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nesprin 2
Polyclonal
Unconjugated
SYNE2
6820443O06Rik; AW546258; D12Ertd777e; EDMD5; KASH domain-containing protein 2; KASH2; KIAA1011; MGC96690; mKIAA1011; Nesp2; Nesp2g; nesprin 2; nesprin-2; NUA; NUANCE; nuclear envelope spectrin repeat protein 2; Nucleus and actin connecting element protein; polytrophin; Protein NUANCE; spectrin repeat containing nuclear envelope protein 2; spectrin repeat containing, nuclear envelope 2; synaptic nuclear envelope 2; synaptic nuclear envelope protein 2; synaptic nuclei expressed gene 2; SYNE2; Syne-2; TROPH
Rabbit
Antigen affinity chromatography
RUO
23224, 319565
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.44 mg/mL
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Q6ZWQ0, Q8WXH0
SYNE2
Recombinant protein encompassing a sequence within the C-terminus region of mouse Nesprin 2. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.