missing translation for 'onlineSavingsMsg'
Learn More

Nesprin 2 Antibody (6C11), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18433497 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433497 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433497 Supplier Novus Biologicals Supplier No. NBP259944MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nesprin 2 Monoclonal antibody specifically detects Nesprin 2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nesprin 2
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6C11
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DKFZP434H2235, DKFZp686E01115, DKFZp686H1931, EDMD5, FLJ46790, KIAA1011FLJ11014, nesprin 2, nesprin-2, NUAFLJ43727, NUANCEFLJ45710, Nuclear envelope spectrin repeat protein 2, Nucleus and actin connecting element protein, polytrophin, Protein NUANCE, spectrin repeat containing, nuclear envelope 2, Synaptic nuclear envelope protein 2, synaptic nuclei expressed gene 2, SYNE-2, TROPH
Host Species Mouse
Immunogen GST fused to residues 6,653-6,835 of human Nesprin 2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 23224
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.