missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nesprin 2 Antibody (10H8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59945
This item is not returnable.
View return policy
Description
Nesprin 2 Monoclonal antibody specifically detects Nesprin 2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Nesprin 2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1-2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
| DKFZP434H2235, DKFZp686E01115, DKFZp686H1931, EDMD5, FLJ46790, KIAA1011FLJ11014, nesprin 2, nesprin-2, NUAFLJ43727, NUANCEFLJ45710, Nuclear envelope spectrin repeat protein 2, Nucleus and actin connecting element protein, polytrophin, Protein NUANCE, spectrin repeat containing, nuclear envelope 2, Synaptic nuclear envelope protein 2, synaptic nuclei expressed gene 2, SYNE-2, TROPH | |
| GST fused to residues 6,653-6,835 of human Nesprin 2 | |
| 0.1 mg | |
| Developmental Biology | |
| 23224 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, Immunofluorescence | |
| 10H8 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction