missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nesprin 1 Monoclonal Antibody (6A)
GREENER_CHOICE

Mouse Monoclonal Antibody

£436.00

Specifications

Antigen Nesprin 1
Clone 6A
Concentration 1 mg/mL
Content And Storage -20°C
Applications Western Blot
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12867772
GREENER_CHOICE
View Documents
Invitrogen™
MA113082
100 μg
£436.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nesprin 1 Monoclonal Antibody for Western Blot

There are two genes encoding members of a new family of type II integral membrane proteins. Both are ubiquitously expressed, and tissue-specific alternative mRNA initiation and splicing generate at least two major isoforms of each protein, with the smaller isoforms being truncated at the N-terminus. These proteins are called nesprin-l and -2 for nuclear _envelope _spectrin repeat, as they are characterized by the presence of multiple, clustered spectrin repeats, bipartite nuclear localization sequences and a conserved C-terminal, single transmembrane domain. Transient transfection of EGFP-fusion expression constructs demonstrated their localization to the nuclear membrane with a novel C-terminal, TM-domain-containing sequence essential for perinuclear localization. Nesprin-l is developmentally regulated in both smooth and skeletal muscle and is relocalized from the nuclear envelope to the nucleus and cytoplasm during C2Cl2 myoblast differentiation. Nesprins may function as 'dystrophins of the nucleus' to maintain nuclear organization and structura integrity.
Specifications

Specifications

Nesprin 1
1 mg/mL
Western Blot
Unconjugated
Mouse
RUO
PBS with 1 mg/mL BSA and 0.05% sodium azide
Q6ZWR6
499010, 64009
Recombinant protein containing residues RKLESTLTGLEQSRERQERR.
Antibody
6A
-20°C
Monoclonal
Liquid
IgG1
Mouse, Rat
SYNE1
8B; A330049M09Rik; ARCA1; BE692247; C130039F11Rik; C6orf98; CPG2; CPG2 full length; dJ45H2.2; DKFZp781J13156; EDMD4; enaptin; enaptin165; FLJ30878; FLJ41140; KASH domain-containing protein 1; KASH1; KIAA0796; KIAA1262; KIAA1756; mKIAA1756; MYNE1; Myne-1; Myocyte nuclear envelope protein 1; Nesp1; nesprin 1; nesprin-1; Nuclear envelope spectrin repeat protein 1; RP1-130E4.2; SCAR8; spectrin repeat containing nuclear envelope protein 1; spectrin repeat containing, nuclear envelope 1; synaptic nuclear envelope 1; synaptic nuclear envelope protein 1; synaptic nuclei expressed gene 1; SYNE 1; SYNE1; Syne-1; syne-1 fragment
SYNE1
Primary
Protein G
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.