missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Nesiritide
Specifications
Specifications
| Purity | 98.45% |
| CAS | 124584-08-3 |
| Content And Storage | Sealed storage, away from moisture and light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light) |
| For Use With (Application) | Cancer-programmed cell death |
| Molecular Weight (g/mol) | 3464.04 g/mol |
| Product Type | Peptides |
| Quantity | 5 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : ≥ 40 mg/mL (11.55 mM) |
| Synonym | Brain Natriuretic Peptide-32 human∣ BNP-32 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?