missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Nesfatin-1/Nucleobindin-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29729
This item is not returnable.
View return policy
Description
Nesfatin-1/Nucleobindin-2 Polyclonal antibody specifically detects Nesfatin-1/Nucleobindin-2 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Nesfatin-1/Nucleobindin-2 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| DNA-binding protein NEFA, NEFAGastric cancer antigen Zg4, nucleobindin 2, nucleobindin-2, nucleobinding 2 | |
| Between 370 and 420 | |
| 100 μg | |
| Signal Transduction | |
| 4925 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction