missing translation for 'onlineSavingsMsg'
Learn More

Nerolidol, cis + trans, 97+%

Product Code. 11473088
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473088 25 g 25g
11483088 100 g 100g
11493088 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473088 Supplier Thermo Scientific Alfa Aesar Supplier No. A18812.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7212-44-4 | C15H26O | 222.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7212-44-4
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.37
MDL Number MFCD00008911,MFCD00008911,MFCD00008911,MFCD00085350
InChI Key FQTLCLSUCSAZDY-SDNWHVSQNA-N
Synonym 3r,6e-nerolidol, unii-uoc0644v25, 3r-6e-nerolidol, nerolidol, 3r,6e-3,7,11-trimethyldodeca-1,6,10-trien-3-ol, e-nerolidol, 1,6,10-dodecatrien-3-ol, 3,7,11-trimethyl-, 3r,6e, nerolidol, 6e--, --nerolidol, ?-nerolidol
PubChem CID 11241545
ChEBI CHEBI:59959
SMILES CC(C)=CCC\C(C)=C\CCC(C)(O)C=C

Specifications

Density 0.876
Boiling Point 263°C to 266°C
Flash Point 96°C (204°F)
Refractive Index 1.479
Quantity 25 g
Beilstein 1724135
Merck Index 14,6477
IUPAC Name (3R,6E)-3,7,11-trimethyldodeca-1,6,10-trien-3-ol
Formula Weight 222.37
Percent Purity ≥97%
Chemical Name or Material Nerolidol, cis + trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.