missing translation for 'onlineSavingsMsg'
Learn More

Nerol, TRC

Product Code. 30472721
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30472721 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30472721

Brand: Toronto Research Chemicals TRCN39009510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nerol
CAS 106-25-2
Molecular Formula C10 H18 O
Quantity 10 g
Synonym 2,6-Octadien-1-ol','3,7-dimethyl-','(2Z)-, 2,6-Octadien-1-ol','3,7-dimethyl-','(Z)- (8CI), Nerol (6CI), (2Z)-3,7-Dimethyl-2,6-octadien-1-ol, (Z)-3,7-Dimethyl-2,6-octadien-1-ol, (Z)-3,7-Dimethyl-2,6-octadienol, (Z)-Geraniol, (Z)-Nerol, 2-cis-3,7-Dimethyl-2,6-octadien-1-ol, 3,7-Dimethyl-cis-2,6-octadien-1-ol, Nerol 900, Neryl alcohol, cis-1-Hydroxy-3,7-dimethyl-2,6-octadiene, cis-3,7-Dimethyl-2,6-octadien-1-ol, cis-Geraniol, β-Nerol
InChI Formula InChI=1S/C10H18O/c1-9(2)5-4-6-10(3)7-8-11/h5,7,11H,4,6,8H2,1-3H3/b10-7-
SMILES CC(=CCC\C(=C/CO)\C)C
IUPAC Name (2Z)-3,7-dimethylocta-2,6-dien-1-ol
Molecular Weight (g/mol) 154.25
Formula Weight 154.1358
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.