missing translation for 'onlineSavingsMsg'
Learn More

Nerol, 97%, Thermo Scientific Chemicals

Product Code. 11458667
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 11458667

Brand: Thermo Scientific Alfa Aesar A19808.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-25-2
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00063204
InChI Key GLZPCOQZEFWAFX-YFHOEESVSA-N
Synonym nerol, cis-geraniol, neryl alcohol, z-3,7-dimethylocta-2,6-dien-1-ol, z-geraniol, 2z-3,7-dimethylocta-2,6-dien-1-ol, 2,6-octadien-1-ol, 3,7-dimethyl-, 2z, nerol natural, cis-3,7-dimethyl-2,6-octadien-1-ol, z-3,7-dimethyl-2,6-octadien-1-ol
PubChem CID 643820
ChEBI CHEBI:29452
IUPAC Name (2Z)-3,7-dimethylocta-2,6-dien-1-ol
SMILES CC(C)=CCC\C(C)=C/CO

Specifications

Density 0.87
Boiling Point 103°C to 105°C (9 mmHg)
Flash Point 77°C (171°F)
Refractive Index 1.475
Quantity 500 mL
Beilstein 1722455
Merck Index 14,6475
Formula Weight 154.25
Percent Purity 97%
Chemical Name or Material Nerol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.