missing translation for 'onlineSavingsMsg'
Learn More

Neratinib, TRC

Product Code. 30468628
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
Product Code. Quantity unitSize
30468628 10 mg 10mg
30465805 100 mg 100mg
30479941 5 mg -
3 options
This item is not returnable. View return policy

Product Code. 30468628

Brand: Toronto Research Chemicals TRCN39009010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698387-09-6
Formula Weight 556.199
InChI Formula InChI=1S/C30H29ClN6O3/c1-4-39-28-16-25-23(15-26(28)36-29(38)9-7-13-37(2)3)30(20(17-32)18-34-25)35-21-10-11-27(24(31)14-21)40-19-22-8-5-6-12-33-22/h5-12,14-16,18H,4,13,19H2,1-3H3,(H,34,35)(H,36,38)/b9-7+
IUPAC Name (E)-N-[4-[3-chloro-4-(pyridin-2-ylmethoxy)anilino]-3-cyano-7-ethoxyquinolin-6-yl]-4-(dimethylamino)but-2-enamide
Molecular Formula C30 H29 Cl N6 O3
Molecular Weight (g/mol) 557.04
SMILES CCOc1cc2ncc(C#N)c(Nc3ccc(OCc4ccccn4)c(Cl)c3)c2cc1NC(=O)\C=C\CN(C)C
Synonym 2-Butenamide, N-[4-[[3-chloro-4-(2-pyridinylmethoxy)phenyl]amino]-3-cyano-7-ethoxy-6-quinolinyl]-4-(dimethylamino)-, (2E)-, (2E)-N-[4-[[3-Chloro-4-(2-pyridinylmethoxy)phenyl]amino]-3-cyano-7-ethoxy-6-quinolinyl]-4-(dimethylamino)-2-butenamide, (E)-N-[4-[3-Chloro-4-[(2-pyridinyl)methoxy]anilino]-3-cyano-7-ethoxy-6-quinolinyl]-4-(dimethylamino)-2-butenamide, (E)-N-[4-[[3-Chloro-4-((pyridin-2-yl)methoxy)phenyl]amino]-3-cyano-7-ethoxyquinolin-6-yl]-4-(dimethylamino)-2-butenamide, HKI 272, Neratinib
Chemical Name or Material Neratinib

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.