missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Fmoc-L-lysine, 97+%

Product Code. p-7041617
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15479298

Brand: Thermo Scientific Alfa Aesar H62834.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 84624-28-2 | C21H24N2O4 | 368.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84624-28-2
Molecular Formula C21H24N2O4
Molecular Weight (g/mol) 368.43
MDL Number MFCD00038365
InChI Key RAQBUPMYCNRBCQ-TWYLJJHKNA-N
Synonym h-lys fmoc-oh, n'-fmoc-l-lysine, s-6-9h-fluoren-9-yl methoxy carbonyl amino-2-aminohexanoic acid, n6-fmoc-l-lysine, lys fmoc-oh, n-, a-fmoc-l-lysine, n-epsilon-9-fluorenylmethyloxycarbonyl-l-lysine, l-lysine, n6-9h-fluoren-9-ylmethoxy carbonyl, 2s-2-amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid
PubChem CID 7010541
IUPAC Name (2S)-2-amino-6-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SMILES N[C@@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Quantity 250 mg
Formula Weight 368.43
Percent Purity 98%
Chemical Name or Material Nepsilon-Fmoc-L-lysine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.