missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine, 95%

Product Code. 15405898
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15405898 2 g 2g
15415898 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15405898 Supplier Thermo Scientific Alfa Aesar Supplier No. H59537.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2389-45-9 | C19H28N2O6 | 380.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2389-45-9
Molecular Formula C19H28N2O6
Molecular Weight (g/mol) 380.44
MDL Number MFCD00065584
InChI Key BDHUTRNYBGWPBL-GGYSOQFKNA-N
Synonym boc-lys z-oh, n-boc-n'-cbz-l-lysine, boc-lys cbz-oh, boc-l-lys z-oh, nalpha-boc-nepsilon-cbz-l-lysine, s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, n-alpha-boc-n-epsilon-benzyloxycarbonyl-l-lysine, nalpha-tert-butoxycarbonyl-nepsilon-carbobenzoxy-l-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-benzyloxycarbonyl-l-lysine
PubChem CID 2724765
SMILES CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Quantity 2 g
Beilstein 1917222
IUPAC Name (2S)-6-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Formula Weight 380.44
Percent Purity 95%
Chemical Name or Material Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.