missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Acetyl-Nalpha-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals

Product Code. 15488718
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488718 5 g 5g
15478718 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488718 Supplier Thermo Scientific Alfa Aesar Supplier No. H62154.06
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159766-56-0
Molecular Formula C23H26N2O5
Molecular Weight (g/mol) 410.47
MDL Number MFCD00077409
InChI Key HQLBYVWJOXITAM-RNZRUAGMNA-N
Synonym fmoc-lys ac-oh, fmoc-n'-acetyl-l-lysine, nalpha-fmoc-nepsilon-acetyl-l-lysine, l-lysine, n6-acetyl-n2-9h-fluoren-9-ylmethoxy carbonyl, 2s-6-acetamido-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, fmoc-lys ac, fmoc-lysine ac-oh, fmoc-l-lys ac-oh, ksc005q1n, fmoc-n-epsilon-acetyl-l-lysine
PubChem CID 7018846
IUPAC Name (2S)-6-acetamido-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SMILES CC(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Quantity 5 g
Formula Weight 410.46
Percent Purity 98%
Chemical Name or Material Nepsilon-Acetyl-Nalpha-Fmoc-L-lysine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.