missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Acetyl-Nalpha-Boc-L-lysine, 98%, Thermo Scientific™

Product Code. 15408878
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408878 250 mg 250mg
15498868 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408878 Supplier Thermo Scientific Alfa Aesar Supplier No. H62347.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6404-26-8
Molecular Formula C13H24N2O5
Molecular Weight (g/mol) 288.34
MDL Number MFCD00057791
InChI Key IOKOUUAPSRCSNT-UEQNJFAPNA-N
Synonym boc-lys ac-oh, n-boc-n'-acetyl-l-lysine, s-6-acetamido-2-tert-butoxycarbonyl amino hexanoic acid, boc-lys-ac-oh, n-boc-n-acetyl-l-lisine, boc-n-epsilon-acetyl-l-lysine, na-t-boc-n-epsilon-acetyl-l-lysine, 2s-6-acetylamino-2-tert-butoxycarbonylamino hexanoic acid, 2s-2-tert-butoxycarbonyl amino-6-acetamidohexanoic acid
PubChem CID 7016049
IUPAC Name (2S)-6-acetamido-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Quantity 250 mg
Formula Weight 288.34
Percent Purity 98%
Chemical Name or Material Nepsilon-Acetyl-Nalpha-Boc-L-lysine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.