missing translation for 'onlineSavingsMsg'
Learn More

Neprilysin/CD10 Antibody (FR4D11), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18350742 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350742 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350742 Supplier Novus Biologicals Supplier No. NBP312068AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Neprilysin/CD10 Monoclonal antibody specifically detects Neprilysin/CD10 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neprilysin/CD10
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone FR4D11
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Atriopeptidase, CALLAmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase), CD10 antigen, CD10), CD10membrane metallo-endopeptidase variant 1, Common acute lymphocytic leukemia antigen, DKFZp686O16152, EC 3.4.24, EC 3.4.24.11, Enkephalinase, EPN, membrane metallo-endopeptidase, membrane metallo-endopeptidase variant 2, MGC126681, MGC126707, NEPmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, neprilysin, Neutral endopeptidase, Neutral endopeptidase 24.11, SFE, Skin fibroblast elastase
Host Species Rabbit
Immunogen This antibody was raised by immunizing mice with Raji cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4311
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.