missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neprilysin-2/MMEL1 Antibody (2D2H5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£435.00
Specifications
| Antigen | Neprilysin-2/MMEL1 |
|---|---|
| Clone | 2D2H5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
Description
Neprilysin-2/MMEL1 Monoclonal specifically detects Neprilysin-2/MMEL1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Neprilysin-2/MMEL1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| EC 3.4.24, EC 3.4.24.11, MELL1, membrane metallo-endopeptidase-like 1, Membrane metallo-endopeptidase-like 2MGC119455, MGC119454, MMEL2, NEP2, NEP2(m), NEPIIMGC119456, Neprilysin II, Neprilysin-2, NL2NL1, SEP, soluble secreted endopeptidase, zinc metallopeptidase | |
| MMEL1 | |
| IgG1 | |
| Protein A or G purified |
| 2D2H5 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 79258 | |
| Purified recombinant fragment of human MMEL1 (AA: 1-107) expressed in E. Coli. | |
| Primary | |
| 89.4 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title