missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nephrin Antibody (G17-H), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP1-30137-0.05ML
This item is not returnable.
View return policy
Description
Nephrin Monoclonal specifically detects Nephrin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
| Nephrin | |
| Monoclonal | |
| 2.5mg/mL | |
| Western Blot 1:2000, ELISA 1:20000-1:50000, Immunoprecipitation 1:10-1:500 | |
| O60500 | |
| NPHS1 | |
| Peptide derived from the region of the first Ig-like domain of human nephrin (between residues Cys53-Cys101). | |
| 0.05 mL | |
| Primary | |
| This antibody is specific for the C terminus of first Ig-like domain of human nephrin (between residues Cys53-Cys101). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunoprecipitation | |
| G17-H | |
| Unconjugated | |
| 20mM Tris/HCl (pH 8.0) and 10 mg/ml BSA with 0.05% Sodium Azide | |
| nephrin, nephrosis 1, congenital, Finnish type (nephrin), NPHNCNF, Renal glomerulus-specific cell adhesion receptor | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4868 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction