missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Nephrin Polyclonal antibody specifically detects Nephrin in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Nephrin |
| Applications | ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | PBS |
| Gene Alias | nephrin, nephrosis 1, congenital, Finnish type (nephrin), NPHNCNF, Renal glomerulus-specific cell adhesion receptor |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a peptide corresponding to 14 amino acids near the carboxy terminus of human Nephrin. The immunogen is located within the last 50 amino acids of Nephrin. Amino Acid Sequence: EPDSLPFELRGHLV |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?