missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nephrin Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-37834B
This item is not returnable.
View return policy
Description
Nephrin Polyclonal antibody specifically detects Nephrin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Nephrin | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 4868 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| nephrin, nephrosis 1, congenital, Finnish type (nephrin), NPHNCNF, Renal glomerulus-specific cell adhesion receptor | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the center of human Nephrin. The immunogen is located within amino acids 870 - 920 of Nephrin. Amino Acid Squence: VPLDLQDPRYTEHTYH | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction