missing translation for 'onlineSavingsMsg'
Learn More

Nepafenac, MedChemExpress

Product Code. 16407256
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16407256 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16407256 Supplier Medchem Express Supplier No. HY17357
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nepafenac(AHR 9434; AL 6515; Nevanac) is a selective COX-2 inhibitor; is prodrug of Amfenac.

Nepafenac(AHR 9434

Specifications

Chemical Name or Material Nepafenac
CAS 78281-72-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H14N2O2
Quantity 100 mg
Synonym AHR 9434 AL 6515
Solubility Information DMSO : 100 mg/mL (393.27 mM; Need ultrasonic)
SMILES O=C(N)CC1=CC=CC(C(C2=CC=CC=C2)=O)=C1N
Molecular Weight (g/mol) 254.28
Formula Weight 254.28
Percent Purity 99.51%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.