missing translation for 'onlineSavingsMsg'
Learn More

Nepafenac-d5, MedChemExpress

Product Code. 16497298
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497298 10 mg 10mg
16469807 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16497298 Supplier Medchem Express Supplier No. HY17357S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nepafenac D5 (AHR-9434 D5) is the deuterium labeled Nepafenac, which is a selective COX-2 inhibitor.

Nepafenac D5 (AHR-9434 D5) is the deuterium labeled Nepafenac, which is a selective COX-2 inhibitor.

Specifications

Chemical Name or Material Nepafenac-d5
CAS 1246814-53-8
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C15H9D5N2O2
Color Yellow
Synonym AHR-9434-d5 AL-6515-d5
Solubility Information DMSO : ≥ 100 mg/mL (385.64 mM)
SMILES O=C(C1=C([2H])C([2H])=C([2H])C([2H])=C1[2H])C2=CC=CC(CC(N)=O)=C2N
Molecular Weight (g/mol) 259.31
Quantity 10 mg
Formula Weight 259.31
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.