missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neostigmine Bromide, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Neostigmine Bromide
Specifications
Specifications
| Chemical Name or Material | Neostigmine Bromide |
| CAS | 114-80-7 |
| Analyte or Component Names | Neostigmine Bromide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H19 N2 O2 . Br |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzenaminium, 3-[[(dimethylamino)carbonyl]oxy]-N,N,N-trimethyl-, bromide, (m-Hydroxyphenyl)trimethylammonium bromide, dimethylcarbamate, Ammonium, (m-hydroxyphenyl)trimethyl-, bromide, dimethylcarbamate, Benzenaminium, 3-[[(dimethylamino)carbonyl]oxy]-N,N,N-trimethyl-, bromide, Neostigmine bromide, Carbamic acid, dimethyl-, ester with (m-hydroxyphenyl)trimethylammonium bromide, (3-Dimethylcarbamoyloxyphenyl)trimethylammonium bromide, Eustigmin bromide, Kirkstigmine bromide, Neoeserine bromide, Neoesserin, Philostigmin bromide, Prostigmin bromide, Prostigmine bromide, Stigmanol bromide, Stigmosan bromide, Syntostigmin (tablet), Syntostigmin bromide, Vagostigmine bromide |
| InChI Formula | InChI=1S/C12H19N2O2.BrH/c1-13(2)12(15)16-11-8-6-7-10(9-11)14(3,4)5;/h6-9H,1-5H3;1H/q+1;/p-1 |
| SMILES | [Br-].CN(C)C(=O)Oc1cccc(c1)[N+](C)(C)C |
| IUPAC Name | [3-(dimethylcarbamoyloxy)phenyl]-trimethylazanium;bromide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction