missing translation for 'onlineSavingsMsg'
Learn More

Neostigmine Bromide, MedChemExpress

Product Code. 16468296
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468296 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468296 Supplier Medchem Express Supplier No. HYB0423
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Neostigmine Bromide is a cholinesterase inhibitor used in the treatment of myasthenia gravis.

Neostigmine Bromide is a cholinesterase inhibitor used in the treatment of myasthenia gravis.

Specifications

Chemical Name or Material Neostigmine Bromide
CAS 114-80-7
Color White
Purity Grade Notes Research
Molecular Formula C12H19BrN2O2
Quantity 100 mg
Synonym Eustigmin bromide Neoserine bromide
Solubility Information DMSO : 100 mg/mL (329.82 mM; Need ultrasonic) ∣H2O : 100 mg/mL (329.82 mM; Need ultrasonic)
SMILES C[N+](C)(C)C1=CC=CC(OC(N(C)C)=O)=C1.[Br-]
Molecular Weight (g/mol) 303.2
Formula Weight 303.2
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.