missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Neoprene O-rings
Description
- Essectial component of a Filtration and Vacuum Desiccator
- Spare O-ring
- Spread a little grease on O-ring for assistance in establishing initial vacuum
Specifications
Specifications
| For Use With (Equipment) | For 8890X |
| Quantity | 1Each |
| Product Type | O-Ring |
| Material | Neoprene |
| Color | Black |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?