missing translation for 'onlineSavingsMsg'
Learn More

Neopentylamine, 97%

Product Code. 15254208 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15254208 25 mL 25mL
15264208 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15254208 Supplier Thermo Scientific Acros Supplier No. 173690250
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5813-64-9 | C5H13N | 87.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5813-64-9
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.17
MDL Number MFCD00008134
InChI Key XDIAMRVROCPPBK-UHFFFAOYSA-N
Synonym neopentylamine, 2,2-dimethylpropylamine, 1-propanamine, 2,2-dimethyl, 2,2-dimethyl-1-propylamine, 1-amino-2,2-dimethylpropane, neo-c5h11nh2, 2,2-dimethyl-1-propylamin, neo-pentylamine, neopentyl amine, 2,2-dimethylpropanamine
PubChem CID 79882
IUPAC Name 2,2-dimethylpropan-1-amine
SMILES CC(C)(C)CN

Specifications

Density 0.7400g/mL
Boiling Point 79.0°C to 82.0°C
Flash Point −13°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)3CCH2NH2
Refractive Index 1.403
Quantity 25 mL
Beilstein 04,188
Specific Gravity 0.74
Solubility Information Solubility in water: soluble.
Formula Weight 87.16
Percent Purity 97%
Chemical Name or Material Neopentylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Neopentylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.