missing translation for 'onlineSavingsMsg'
Learn More

Neopentyl bromide, 98%

Product Code. 11352226
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352226 5 g 5g
11362226 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352226 Supplier Thermo Scientific Alfa Aesar Supplier No. L03579.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 630-17-1 | C5H11Br | 151.05 g/mol

Neopentyl bromide is used in synthesis of S-alkylated cysteine derivatives with branched alkyl chains. It can be used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Neopentyl bromide is used in synthesis of S-alkylated cysteine derivatives with branched alkyl chains. It can be used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Soluble in ethanol, ether, aetone and benzene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-17-1
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.05
MDL Number MFCD00000209
InChI Key CQWYAXCOVZKLHY-UHFFFAOYSA-N
Synonym neopentyl bromide, propane, 1-bromo-2,2-dimethyl, 1-bromo-2,2-dimethyl-propane, ccris 4895, neopentylbromde, neopentylbromide, neo-pentyl bromide, ch3 3cch2br, 3-bromo-2,2-dimethylpropyl, neopentyl bromide 5g
PubChem CID 12415
IUPAC Name 1-bromo-2,2-dimethylpropane
SMILES CC(C)(C)CBr

Specifications

Density 1.195
Boiling Point 104°C to 106°C
Flash Point 6°C (42°F)
Refractive Index 1.436
Quantity 5 g
UN Number UN1993
Beilstein 1730989
Solubility Information Soluble in ethanol,ether,aetone and benzene. Insoluble in water.
Formula Weight 151.05
Percent Purity 98%
Chemical Name or Material Neopentyl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.