Learn More
Invitrogen™ Neon™ NxT Electroporation System Tubes
Description
Neon NxT Electroporation System Tubes are designed specifically for use with the Neon NxT Electroporation System (Cat. No. NEON1). Each Neon NxT Electroporation System Kit for 25 x 2 reactions (Cat. Nos. N1025 and N10025) comes with eight electroporation tubes and each Neon NxT Electroporation System Kit for 96 x 2 reactions (Cat. Nos. N1096 and N10096) comes with 32 electroporation tubes. This pack of 96 tubes is offered for those wishing to use single tubes per reaction depending on their experimental setup.
Specifications
Specifications
| Cell Type | Stem Cells, Primary Cells, Hard-to-Transfect Cells |
| Format | Electroporation Pipette Tip |
| Transfection Technique | Electroporation |
| For Use With (Equipment) | Neon™ NxT Electroporation System |
| For Use With (Application) | Transfection |
| Quantity | 96 tubes |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.