Learn More
Invitrogen™ Neon™ NxT Electroporation System 8-Channel Tubes
Product Code. 30245026
Change view
Click to view available options
Quantity:
8 Tubes
Unit Size:
Pack of 8
Product Code. Quantity Unit Size
30245026 8 Tubes Pack of 8
1 options
Product Code. 30245026 Supplier Invitrogen™ Supplier No. NT8
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Neon NxT Electroporation System 8-Channel Tubes are designed specifically for use with the Neon NxT Electroporation System with 8-Channel Pipette.
Neon NxT Electroporation System 8-Channel Tubes are designed specifically for use with the Neon NxT Electroporation System with 8-Channel Pipette (Cat. Nos. NEON18S, NEON18SK, NEON8, NEON8U). Each Neon NxT Electroporation System Kit with 8-Channel Tubes for 96 x 2 reactions (Cat. Nos. N1096-8 and N10096-8) comes with four 8-channel tubes and each Neon NxT Electroporation System Kit for 384 x 2 reactions (Cat. Nos. N10384-8 and N100384-8) comes with sixteen 8-channel tubes. This pack of eight tubes is offered for those wishing to use one tube per eight reactions (tubes can be used up to twelve times) depending on their experimental setup.
TRUSTED_SUSTAINABILITY
Specifications
Includes Includes 8 x 8-chamber tubes, use at room temperature under normal laboratory conditions. Refer to the Neon™ NxT Electroporation System user manual for protocol instructions.
Quantity 8 Tubes
For Use With (Equipment) Neon NxT Electroporation System
For Use With (Application) Electroporation