missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Neon™ NxT Electroporation System 100-μL Kit with 1-Channel Tubes

Product Code. p-7229402
Change view
Click to view available options
No. of Reactions:
25 x 2 Reactions
96 x 2 Reactions
Unit Size:
Each
Pack of 25
Pack of 96
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
17809444 25 x 2 Reactions Each
18035331 25 x 2 Reactions Pack of 25
17819444 96 x 2 Reactions Each
18015341 96 x 2 Reactions Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17809444 Supplier Invitrogen™ Supplier No. N10025

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Neon NxT Electroporation System 100-μL Kit is designed specifically for use with the Neon NxT Electroporation System.

The Neon NxT Electroporation System 100-μL Kit with 1-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System (Cat. Nos. NEON1S, NEON1SK, NEON18S, NEON18SK). Use this kit for transfection volumes of 100 μL, containing 5 x 105–2 x 106 adherent cells or 1 x 106–5 x 106 suspension cells. Cells that have been transfected using the included 100-μL Neon NxT tips are then ready to be washed and plated into a multi-well culture dish.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 10 mL Resuspension R Buffer; store at 4°C
• 10 mL Resuspension T Buffer; store at 4°C
• 10 mL Resuspension Genome Editing Buffer; store at 4°C
• 50 mL Electrolytic E100 Buffer; store at 4°C
• 25 reaction tips
• 8 electroporation 1-channel tubes
Cell Type Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format Electroporation Pipette Tip
Transfection Technique Electroporation
For Use With (Equipment) Neon™ NxT Electroporation System
For Use With (Application) Transfection
No. of Reactions 25 x 2 Reactions
Product Type Pipette Tip
Quantity 25 x 2 reactions
Volume (Metric) Sample 100 μL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.