missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Neon™ NxT Electroporation System 10 μL Kit with 8-Channel Tubes

Product Code. p-200087435
Change view
Click to view available options
No. of Reactions:
384 x 2 Reactions
96 x 2 Reactions
Unit Size:
1 set
Product Code. No. of Reactions unitSize
30245458 96 x 2 Reactions 1 set
30245218 384 x 2 Reactions 1 set
2 options
This item is not returnable. View return policy

Product Code. 30245458

Brand: Invitrogen™ N10968

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Neon NxT Electroporation System 10-μL Kit with 8-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System with 8-channel pipette and pipette station.

The Neon NxT Electroporation System 10-μL Kit with 8-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System (Cat. No. MPK5000) with 8-channel pipette and pipette station (Cat. Nos. NEON18S, NEON18SK, NEON8, NEON8U). Use this kit for transfection volumes of 10 μL, containing 5 x 104 to 2 x 105 adherent cells or 1 x 105 to 5 x 105 suspension cells. Cells that have been transfected using the included 10-μL Neon NxT tips are ready to be plated into a multi-well culture dish.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Stem Cell, Primary Cell, Hard-to-Transfect Cell
Content And Storage Box 1 includes:
• Resuspension R Buffer, 4 x 1 mL
• Resuspension T Buffer, 4 x 1 mL
• Resuspension Genome Editing Buffer, 4 x 1 mL
• Electrolytic E10 Buffer, 2 x 100 mL

Store at 4°C.

Box 2 includes:
• Reaction tips, 96 tips
• Electroporation 8-channel tubes, 4 tubes

Store at room temperature.
Format Electroporation Pipette Tip
Transfection Technique Electroporation
For Use With (Equipment) Neon™ NxT Electroporation System
For Use With (Application) Transfection
No. of Reactions 96 x 2 Reactions
Quantity 96 x 2 Reactions
Volume (Metric) Sample 10 μL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.