Learn More
Invitrogen™ Neon™ NxT Electroporation System 10 μL Kit with 8-Channel Tubes
Description
The Neon NxT Electroporation System 10-μL Kit with 8-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System with 8-channel pipette and pipette station (Cat. Nos. NEON18S, NEON18SK, NEON8, NEON8U). Use this kit for transfection volumes of 10 μL, containing 5 x 104 to 2 x 105 adherent cells or 1 x 105 to 5 x 105 suspension cells. Cells that have been transfected using the included 10-μL Neon NxT tips are ready to be plated into a multi-well culture dish.
Specifications
Specifications
| Cell Line | Stem Cell, Primary Cell, Hard-to-Transfect Cell |
| Content And Storage | Box 1 includes: • Resuspension R Buffer, 4 x 1 mL • Resuspension T Buffer, 4 x 1 mL • Resuspension Genome Editing Buffer, 4 x 1 mL • Electrolytic E10 Buffer, 2 x 100 mL Store at 4°C. Box 2 includes: • Reaction tips, 96 tips • Electroporation 8-channel tubes, 4 tubes Store at room temperature. |
| Format | Electroporation Pipette Tip |
| Transfection Technique | Electroporation |
| For Use With (Equipment) | Neon™ NxT Electroporation System |
| For Use With (Application) | Transfection |
| No. of Reactions | 96 x 2 Reactions |
| Quantity | 96 x 2 Reactions |
| Volume (Metric) Sample | 10 μL |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.