Description
Neomycin sulfate is an aminoglycoside antibiotic active against gram-negative bacteria. It is commonly used in many topical medications, such as creams, ointments, and eyedrops. It binds to duplex RNA with high affinity and inhibits protein biosynthesis. Neomycin sulfate also acts as a calcium channel blocker. Usage Statement: Unless specified otherwise, MP Biomedical™ products are for research or further manufacturing use only, not for direct human use.
Specifications
Specifications
| Physical Form | Powder |
| Color | White |
| Quantity | 10 g |
Safety and Handling
- Neomycin Sulfate, Cell Culture Reagent
- Danger
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-[In case of inadequate ventilation] wear respiratory protection.
- P321-Specific treatment (see on this label).
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Neomycin Sulfate