missing translation for 'onlineSavingsMsg'
Learn More

Neomycin Sulfate, MP Biomedicals™
BIOPHARMA

Product Code. p-7083050 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 17650887

Brand: MP Biomedicals 0218061005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acts against both gram-positive and gram-negative bacteria. Neomycin Sulfate, MP Biomedicals is produced by Streptomyces containing a minimum of 85% neomycin B.

This product acts by binding to the 30S and 50S subunits, causing miscoding and inhibiting initiation and elongation during protein synthesis. Neomycin also blocks voltage-sensitive Ca2+ channels without affecting the Na+/Ca2+ antiporter in neurons.

Applications:

  • As a selection agent for prokaryotic cells transformed using the neo selectable marker gene
  • To study ototoxic side effects of antibiotics and platelet-derived growth factor responses in certain fibroblasts
  • In the extraction of Nuclear Phosphatidylinositol 4,5-Bisphosphate-Interacting Proteins
  • It is recommended for use in cell culture applications at 50mg/L
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-10-3
Molecular Formula C23H48N6O17S
Molecular Weight (g/mol) 712.72
MDL Number MFCD29905465
InChI Key OIXVKQDWLFHVGR-DJUBFIBWNA-N
PubChem CID 124080932
SMILES OS(O)(=O)=O.NCC1OC(O[C@@H]2[C@@H](CO)O[C@@H](OC3C(O)C(N)CC(N)C3OC3OC(CN)C(O)C(O)C3N)[C@@H]2O)C(N)C(O)C1O

Specifications

Physical Form Powder
pH 5 to 7.5
Loss on Drying <8.0%
Quantity 5 g
Solubility Information H2O soluble (50mg/mL)
IUPAC Name 5-amino-2-(aminomethyl)-6-{[(2R,3S,4R,5S)-5-[(3,5-diamino-2-{[3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy}-6-hydroxycyclohexyl)oxy]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy}oxane-3,4-diol; sulfuric acid
Formula Weight 908.9g/mol
Grade USP
compliance-icons
Product Identifier
  • Neomycin Sulfate, USP
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P321-Specific treatment (see on this label).
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Neomycin Sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.